Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10630

Sigma-Aldrich

AFB-Color malachite green (oxalate) solution

for the microscopic inverstigation of acid-fast bacteria (cold staining)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

liquid

IVD

for in vitro diagnostic use

density

1.0 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

AFB-Color malachite green (oxalate) solution - for the microscopic investigation of acid-fast bacteria (AFB) (cold staining), is a ready-to-use solution used for human-medical cell diagnosis and serves the purpose of the bacteriological investigation of sample material of human origin., for example smears of enriched bacterial cultures. Acid-fast bacteria are difficult to stain because of the high proportion of lipid and wax in their cell walls. The most efficient staining method is the hot staining according to Ziehl-Neelsen. In this method, carbolfuchsin solution is applied to the specimen and heated. This heating process accelerates the rate at which the fuchsin dye is absorbed and thus the formation of the mycolate-fuchsin complex in the cell wall. In this case, a modification of the AFB-Color carbol fuchsin (Product number 1.08512) solution renders heating unnecessary (Kinyoun method, cold staining). Once the acid-fast bacteria have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol. Accordingly, acid-fast bacteria are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye. In this instruction for use malachite green is used for counterstaining.
Pretreatment of the specimens with Sputofluol (Product number 1.08000) dissolves the bacteria from the surrounding viscid sputum and cell material.
The 500 ml bottle of AFB-Color malachite green (oxalate) solution provides 250 stainings. It is an IVD product and CE registered. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Used to detect mycobacteria (AFB cold staining)

Analysis Note

Suitability for microscopypasses testMycobacteriaredBackgroundlight green

Legal Information

SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service