Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10449

Millipore

DNase test agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

powder (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Staphylococcus spp.

General description

for microbiology

Analysis Note

Appearance
- clearness:
clear to slightly opalescent
- colour: yellowish brown
pH-value (25 °C): 7.1 - 7.5
Growth
- Staphylococcus aureus ATCC 25923 (WDCM 00034):
good to very good
- Staphylococcus aureus ATCC 6538-P (WDCM 00033): good to very good
- Staphylococcus epidermidis ATCC 12228 (WDCM 00036): good to very good
- Escherichia coli ATCC 25922 (WDCM 00013): good to very good
- Serratia marcescens ATCC 14756: good to very good
- Bacillus cereus ATCC 11778 (WDCM 00001): good to very good
Clear zones
- Staphylococcus aureus ATCC 25923 (WDCM 00034):
+
- Staphylococcus aureus ATCC 6538-P (WDCM 00033): +
- Staphylococcus epidermidis ATCC 12228 (WDCM 00036): -
- Escherichia coli ATCC 25922 (WDCM 00013): -
- Serratia marcescens ATCC 14756: +
- Bacillus cereus ATCC 11778 (WDCM 00001): +
Incubation: 24 hrs.; 35 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service