Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09993

Supelco

Strontium standard

1000 mg Sr, (SrCl₂ in 7% HCl) Titrisol®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

Quality Level

product line

Titrisol®

form

concentrate

technique(s)

AAS: suitable
ICP: suitable

pH

1 (20 °C in H2O)

density

1.06 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

content (Sr): 1000 mg / Ampoule
Determination method: Complexometric titration.
(traceable of NIST - SRM 682)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service