Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09960

Supelco

Cadmium standard

1000 mg Cd, (CdCl₂ in H₂O) Titrisol®

Synonym(s):

Cadmium standard for aqueous matrices

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Cd in H2O

technique(s)

AAS: suitable
ICP: suitable

pH

4.2 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Analysis Note

content (Cd): 1000 mg / Ampoule
Determination method:Complexometric titration.
(traceable of NIST - SRM 682)
Accuracy of the method:+/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service