Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09869

Supelco

Fluoride standard

1000 mg F (KF in H₂O) Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$86.90

$86.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 AMP
$86.90

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$86.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Titrisol®

technique(s)

ion chromatography: suitable

pH

8.4 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Analysis Note

content (F⁻): 1000 mg / Ampoule
Determination method:Titration with Lanthannitrat solution (traceable to NIST SRM 682)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service