Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.09798

Supelco

o-Xylene

reference substance for gas chromatography

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-422-2

vapor density

3.7 (vs air)

Quality Level

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

867 °F

expl. lim.

7 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

transition temp

flash point 30 °C

solubility

0.18 g/L

density

0.879 g/mL at 20 °C (lit.)

storage temp.

2-30°C

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • o-Xylene for spectroscopy and chromatography: The use of o-Xylene in the field analysis of gas-phase VOCs highlights its application as an effective solvent in compound-specific isotope analysis, underlining its importance in environmental monitoring and pharmaceutical research (J Contam Hydrol, 2024; doi: 10.1016/j.jconhyd.2024.104310).

  • o-Xylene in molecular biology research applications: Research utilizes o-Xylene in molecular dynamics simulations to understand the adsorption and diffusion behaviors of aromatic compounds, providing insights crucial for the development of advanced materials and pharmaceutical applications (Langmuir, 2024; doi: 10.1021/acs.langmuir.3c03706).

Analysis Note

Assay (GC): ≥ 99.0 %
Identity (IR-spectrum): conforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service