Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09716

Supelco

n-Octane

reference substance for gas chromatography

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-892-1

vapor pressure

14 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

210 °C

expl. lim.

0.8-6.5 % (v/v)

technique(s)

gas chromatography (GC): suitable

bp

125-126 °C/1013 hPa

mp

-57 °C

transition temp

flash point 13 °C

density

0.70 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service