1.09711
Hydrazine Test, photometric
photometric, 0.005-2.00 mg/L (N2H4), Spectroquant®
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Hydrazine Test, photometric, 0.005-2.00 mg/L (N2H4), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 100 tests
specific analyte(s)
hydrazine
measuring range
0.005-2.00 mg/L (N2H4)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (4-(Dimethylamino)-benzaldehyde)
storage temp.
15-25°C
General description
The method is analogous to DIN 38413-1.
This Spectroquant® Hydrazine Reagent Test allows the accurate quantification of the hydrazine content in aqueous samples.
Method applied: In acidic solution hydrazine reacts with 4-(dimethylamino) benzaldehyde to form a yellow compound which is determined photometrically.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1 - Skin Sens. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service