Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.09511

Supelco

Lead(II)acetate paper

roll L 4.8 m, MQuant®

Synonym(s):

Lead(II)acetate paper chemical test strips, Lead(II)acetate paper test paper

Sign Into View Organizational & Contract Pricing

Select a Size

3 ROLLS
$73.10

$73.10


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
3 ROLLS
$73.10

About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

$73.10


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

description

semi-quantitative

Quality Level

product line

MQuant®

form

solid

specific analyte(s)

sulfide

packaging

pkg of 3 rolls

roll L

4.8 m

detection method

colorimetric

storage temp.

15-25°C

General description

roll length 4.8 m

Lead (II) acetate paper is used for the determination of sulfide and hydrogen sulfide. In contact with sulfide, it forms brownish black lead sulfide. This is a qualitative test paper, telling whether sulfide is in the sample or not. For a semiquantitative or quantitative result, please consider a test from our MQuant® or Spectroquant® portfolio.

Analysis Note

Application test: conforms

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I need the using process of "1.09511.0003 Lead (II) acetate paper roll length 4.8m". For example: put the paper to contact the teste gas is enough or not. or I need to put it into the liquid?

    1 answer
    1. Lead(II) acetate paper can be utilized to measure both gas and liquid. The paper can be directly placed in the liquid or a moistened paper (with distilled water) can be exposed to the gas. The time needed for the reaction depends on the sulfide concentration, but a color change will be visible within one minute at the latest.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service