Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09122

Supelco

Potassium permanganate solution

c(KMnO4) = 0.02 mol/L (0.1N), standardized with oxalate, reag. USP, ready-to-use for titration, Titriplex®

Synonym(s):

KMnO4 Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352312
NACRES:
NA.21

product name

Potassium permanganate solution, standardised against oxalate c(KMnO4) = 0.02 mol/l (0.1 N), Titripur®, reag. USP

Agency

reag. USP

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Redox Reactions

concentration

0.02 M

technique(s)

titration: suitable

pH

8 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Oxidation of chromium(III): A potential risk of using chemical oxidation processes for the remediation of 2-chlorophenol contaminated soils: This research highlights the oxidation of chromium using potassium permanganate, emphasizing the environmental risks and the need for controlled application in soil remediation efforts (Yu et al., 2024).

  • Potential in-vitro antioxidant and anti-inflammatory activity of Martynia annua extract mediated Phytosynthesis of MnO(2) nanoparticles: Discusses the use of potassium permanganate in the synthesis of manganese dioxide nanoparticles, exploring their potential biomedical applications in antioxidant and anti-inflammatory treatments (Thangapushbam et al., 2024).

  • Investigating the impact of external application of formalin and potassium permanganate on hematological, immunological, and biochemical profiles in Labeo rohita fingerlings: Examines the effects of potassium permanganate treatments on fish health, providing insights into its safe usage in aquaculture (Banjare et al., 2024).

  • Development of a thin layer chromatography-based method to detect sorbitol presence in milk and its applicability in formalin preserved milk samples: This study utilizes potassium permanganate in developing a new analytical method for detecting additives in dairy products, demonstrating its utility in food safety and quality assurance (Kumar et al., 2024).

  • Field and laboratory studies of fluorescence-based technologies for real-time tracking of cyanobacterial cell lysis and potential microcystins release: Research on the application of potassium permanganate in managing cyanobacterial blooms and associated toxin release in water bodies, offering advancements in environmental monitoring and public health protection (Tsai et al., 2024).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Linkage

Replaces: PX1553-1

Analysis Note

Form liquid
Amount-of-substance concentration 0.01990 - 0.02010 mol/L
Measurement uncertainty ± 0.00006 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 8040 di-sodium oxalate) by means of volumetric standard di-sodium oxalate (article number 1.02407), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Hydrogen peroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service