Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08828

Supelco

Zinc iodide

for analysis EMSURE®

Synonym(s):

Zinc iodide, Zinc diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$346.00

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$346.00

About This Item

Empirical Formula (Hill Notation):
I2Zn
CAS Number:
Molecular Weight:
319.20
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-396-0
NACRES:
NA.21
form:
solid
solubility:
4500 g/L

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥98.5% (complexometric)

form

solid

impurities

≤0.00005% As (Arsenic)
≤0.001% Cu (Copper)
≤0.001% Fe (Iron)
≤0.001% Pb (Lead)
≤0.005% Cd (Cadmium)
≤0.005% Sulfate (SO4)
≤0.01% Chloride (Cl)
≤0.1% Substances not precipitated by ammonium sulfide (as sulfates)

loss

≤1.5% loss on drying

pH

5.6 (20 °C, 50 g/L in H2O)

bp

625 °C/1013 hPa

mp

446 °C

solubility

4500 g/L

density

4.74 g/cm3 at 20 °C

bulk density

1800 kg/m3

storage temp.

2-30°C

SMILES string

[Zn+2].[I-].[I-]

InChI

1S/2HI.Zn/h2*1H;/q;;+2/p-2

InChI key

UAYWVJHJZHQCIE-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): ≥ 98.5 %
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.005 %
As (Arsenic): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Pb (Lead): ≤ 0.001 %
Substances not precipitated by ammonium sulfide (as sulfates): ≤ 0.1 %
Loss on drying: ≤ 1.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service