Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08602

Millipore

N-[Tris(hydroxymethyl)methyl]glycine

buffer substance TRICIN

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine, Tricine, N-Tris(hydroxymethyl)methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12161703
EC Index Number:
227-193-6
NACRES:
NA.05
Pricing and availability is not currently available.

Quality Level

pH

4.5-5.5 (20 °C, 50 g/L in H2O)

mp

187 °C

solubility

100 g/L

bulk density

500 kg/m3

storage temp.

15-25°C

SMILES string

[N+H2](C(CO)(CO)CO)CC(=O)[O-]

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

General description

buffer substance TRICIN

Analysis Note

Assay (perchloric acid titration)≥ 99 %Identity (IR-spectrum)passes testUV-Absorption (250 nm, 50 g/l, 1 cm, water)≤ 0.075Chloride (Cl)≤ 0.05 %Sulphate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Na (Sodium)≤ 0.005 %Loss on Drying (105°C)≤ 0.2 %Biological buffer, pKa at 20 °C8.15

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service