Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.08498

Supelco

Vanadate-molybdate reagent

for determination of phosphate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

vapor pressure

<20 hPa ( 20 °C)

Quality Level

form

liquid

specific analyte(s)

phosphate

technique(s)

photometry: suitable

pH

<1 (20 °C, 50 g/L in H2O)

bp

>100 °C/1013 hPa

mp

<-4 °C

density

1.15 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Sulfuric acid (alkalimetric): 18.0 - 20.5 %
Application test: conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service