Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.08440

Supelco

1,1,2-Trichlorotrifluoroethane

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

1,1,2-Trichlorotrifluoroethane, 1,1,2-Trifluorotrichloroethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.37
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-936-1
grade:
reagent grade
bp:
47.6 °C/1013 hPa
vapor pressure:
360 hPa ( 20 °C)

grade

reagent grade

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

360 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

680 °C

potency

43000 mg/kg LD50, oral (Rat)

dilution

(for analytical testing)

impurities

≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000002% Cu (Copper)
≤0.000002% Mn (Manganese)
≤0.000002% Ni (Nickel)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00001% Fe (Iron)
≤0.00001% Mg (Magnesium)
≤0.00001% Pb (Lead)
≤0.00001% Sn (Tin)
≤0.00001% Zn (Zinc)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.0002% Iodine reducing matter (as HCHO)
≤0.0005% Free acid (as HCl)
≤0.005% Water
≤0.01% Carbon tetrachloride (GC)
≤0.01% Chloroform (GC)
≤0.01% Dichloromethane (GC)
≤3050-2900 cm⁻¹)) ≤ 5 ppm squalene

color

APHA: ≤10

bp

47.6 °C/1013 hPa

mp

-35 °C

transition temp

flash point 195 °C

solubility

0.17 g/L

density

1.58 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

General description

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
for analysis EMSURE® Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(C)

195 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service