Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.08435

Supelco

Titriplex® VI

for analysis

Synonym(s):

Titriplex® VI, 3,6-Dioxaoctamethylenedinitrilotetraacetic acid, Ethylenedioxybis(ethylenenitrilo)tetraacetic acid, Bis(aminoethyl)glycolether-N,N,N,N′-tetraacetic acid, Ethylene glycol-bis (ß-amino ethyl ether)-N,N′-tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N2O10
CAS Number:
Molecular Weight:
380.35
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-651-2
NACRES:
NA.21

Quality Level

form

solid

potency

3587 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

mp

240-245 °C (decomposes)

bulk density

450 kg/m3

storage temp.

15-25°C

InChI

1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

DEFVIWRASFVYLL-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 98.5 %
Identity (IR-spectrum): conforms
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): ≤ 1.0 %

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service