Skip to Content
MilliporeSigma

1.08432

SAFC

Urea

crystalline, EMPROVE® API, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

Carbamide, Carbonyl diamide, Carbonyldiamine, Diaminomethanone, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-315-5
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

GMO free
GMP
Halal
Kosher

Agency

BP
Ph. Eur.
USP

product line

EMPROVE® API

form

crystalline

impurities

low in endotoxins

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

application(s)

ophthalmics
pharmaceutical
semi-solid formulation

foreign activity

Aflatoxin free

storage temp.

2-25°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Urea as an active pharmaceutical ingedient is used mainly in dermatology for cremes and ointments and for ophthalmic applications. Urea crystalline Emprove® API is manufactured under ICH Q7. A CEP and an US-DMF is available.
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Packaging

1.08432.0001 // 3x 1kg // PE bottle
1.08432.1000 // 1kg // PE bottle
1.08432.5000 // 5kg // PE bottle
1.08432.9029 // 25 kg //Double PE bag

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Elemental Impurities Information

Available by subscription or for a fee

API Information Package

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service