Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.08239

Supelco

1,1,2-Trichlorotrifluoroethane

for IR spectroscopy Uvasol®

Synonym(s):

1,1,2-Trichlorotrifluoroethane, 1,1,2-Trifluorotrichloroethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Cl3F3
CAS Number:
Molecular Weight:
187.38
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-936-1

vapor pressure

360 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

680 °C

potency

43000 mg/kg LD50, oral (Rat)

technique(s)

IR spectroscopy: suitable
UV/Vis spectroscopy: suitable

bp

47.6 °C/1013 hPa

mp

-35 °C

transition temp

flash point 195 °C

solubility

0.17 g/L

density

1.58 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

383.0 °F

Flash Point(C)

195 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service