Skip to Content
MilliporeSigma

1.08212

SAFC

Parteck® PLX 188 (Poloxamer 188)

(stabilized with 70ppm BHT), EMPROVE® ESSENTIAL, Ph. Eur., NF

Pharma Manufacturing

Synonym(s):

Poloxamer 188

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H6O · C2H4O)x
CAS Number:
MDL number:
UNSPSC Code:
12352112

biological source

synthetic

Quality Level

Agency

NF
Ph. Eur.

product line

EMPROVE® ESSENTIAL

bp

>150 °C

mp

55 °C

bulk density

1050 kg/m3

storage temp.

2-25°C

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid oral dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® PLX 188 excipient combines adequate lubrication at tolerable concentrations with the added benefit of dissolution enhancement. It features consistent performance, improved dissolution rate and reliable lubrication – all at the same time. As a result, Parteck® PLX 188 excipient can therefore provide a simple solution to low bioavailability of DCS 2a compounds.

Features and Benefits

  • Hydrophilic, water-soluble tablet lubrication
  • Improved dissolution rates for poorly soluble DCS class IIa molecules
  • Specified particle size distribution optimized for solid application performance
  • Regulatory support through compliance with Ph. Eur. and USP-NF, going beyond current requirements with specitied formaldehyde content

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our overview of API solubility and dissolution enhancement methods can help you decide between the many options to take during formulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service