Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08175

Supelco

Thymolphthalein

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Thymolphthalein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$132.00
25 G
$330.00
100 G
$1,150.00

$132.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$132.00
25 G
$330.00
100 G
$1,150.00

About This Item

Empirical Formula (Hill Notation):
C28H30O4
CAS Number:
Molecular Weight:
430.54
MDL number:
UNSPSC Code:
12171502
EC Index Number:
204-729-7
NACRES:
NB.21

$132.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Thymolphthalein, indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

loss

≤1% loss on drying, 110°C

color

white to almost white

visual transition interval

9.0-10.5, colorless to blue

mp

253 °C

bulk density

400 kg/m3

λmax

592-596 nm (buffer pH 10.5)

storage temp.

2-30°C

SMILES string

O1C(c4c(cccc4)C1=O)(c3c(cc(c(c3)C(C)C)O)C)c2c(cc(c(c2)C(C)C)O)C

InChI

1S/C28H30O4/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(31)32-28)24-14-21(16(3)4)26(30)12-18(24)6/h7-16,29-30H,1-6H3

InChI key

LDKDGDIWEUUXSH-UHFFFAOYSA-N

Application


  • Electrode Based on the MWCNTs and Electropolymerized Thymolphthalein for the Voltammetric Determination of Total Isopropylmethylphenols in Spices: This study developed a sensitive electrochemical sensor for quantifying thymol and carvacrol in spices using an electrode modified with multi-walled carbon nanotubes and electropolymerized thymolphthalein (Chernousova et al., 2023).

  • Fluorescence Enhancement Mechanism of Thymolphthalein-Based Probe by Coordination Interaction with Zinc Ion: This research explored the fluorescence enhancement of a thymolphthalein-based probe through coordination with zinc ions, highlighting its potential in analytical applications (Huang et al., 2021).

  • pH-Triggered Visual Detection of Escherichia coli Based on the Co-Assembly of Bacitracin and Thymolphthalein: This article presents a novel method for detecting E. coli through pH-triggered visual changes using a thymolphthalein and bacitracin-based probe (Lin et al., 2023).

  • Bio-Thymol Containing New High-Performance Thymolphthalein-Based Polybenzoxazine: Thermal, Superhydrophobic and Dielectric Properties: The study reports on a new polybenzoxazine material incorporating thymolphthalein, showcasing enhanced thermal, hydrophobic, and dielectric properties (Srinivasan et al., 2023).

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Appearance: White to almost white powder, eventually slightly yellowish.
Clarity of solution (1 g/l; ethanol): passes test
Absorption maximum λmax. (buffer pH 10.5): 592 - 596 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; buffer pH 10.5; calculated on anhydrous substance): 800 - 900
Transition range: pH 9.0 - pH 10.5 colourless - blue
Transition range (according to ACS): passes test
Transition interval (according Reag. Ph. Eur.): passes test
Loss on drying (110 °C): ≤ 1 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service