Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.07749

Supelco

Silica gel 60 PF₂₅₄ containing gypsum

for preparative layer chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
NACRES:
NB.21

material

silica gel 60 matrix

Quality Level

form

powder

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 2.5 kg

technique(s)

preparative TLC: suitable
thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

300 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Linkage

Replaces: 7749-3

Analysis Note

pH-value (10 % suspension): 6.0 - 7.0
Calcium sulfate (CaSO₄·0.5 H₂O): 28 - 32 %
Fe (Iron): ≤ 0.02 %
Particle size: 90% ≤55 µm
Application test: conforms
The data to the particle size are related to the pure silica gel before additive addition.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service