Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.07463

Supelco

Pyridine

dried (max. 0.0075% H₂O) SeccoSolv

Synonym(s):

Pyridine, Azine, Azabenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-809-9

vapor pressure

26.7 hPa ( 25 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

482 °C

potency

891 mg/kg LD50, oral (Rat)
1121 mg/kg LD50, skin (Rabbit)

technique(s)

DNA amplification: suitable

impurities

≤0.0075% Water
≤0.01% Piperidine (GC)
≤0.2% 2-Picoline (GC)

evapn. residue

≤10 mg/L

color

APHA: ≤10

pH

8.81 (20 °C in H2O)

bp

115 °C/1013 hPa

mp

-42 °C

transition temp

flash point 17 °C

solubility

soluble 1000 g/L

density

0.982 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

2-30°C

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Analysis Note

Evaporation Number: 12.7
Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Color: ≤ 10 Hazen
2-Picoline (GC): ≤ 0.2 %
Piperidine (GC): ≤ 0.01 %
Reducing substances: conforms
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.0075 %

Legal Information

SECCOSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service