Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07055

Supelco

Petroleum benzine

for analysis EMPARTA® ACS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12191502
grade:
ACS reagent

grade

ACS reagent

dilution

(for routine analytical applications)

storage temp.

20-25°C

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<-5.8 °F

Flash Point(C)

< -21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service