Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07046

Supelco

Dimethyl sulfoxide

for analysis EMPARTA® ACS

Synonym(s):

Dimethyl sulfoxide, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
12191502
grade:
ACS reagent
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

dilution

(for routine analytical applications)

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

storage temp.

20-25°C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service