Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06509

Supelco

Sodium thiosulfate pentahydrate

99.999 Suprapur®, for inorganic trace analysis

Synonym(s):

Sodium thiosulfate pentahydrate, Antichlor

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$469.00

$469.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$469.00

About This Item

Empirical Formula (Hill Notation):
Na2O3S2 · 5H2O
CAS Number:
Molecular Weight:
248.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-867-5
NACRES:
NA.21
grade:
for inorganic trace analysis
form:
solid
solubility:
701 g/L

$469.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for inorganic trace analysis

Quality Level

Assay

99.999%

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

mp

48 °C

solubility

701 g/L

density

1.74 g/cm3 at 20 °C

bulk density

1000 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=S)(=O)([O-])[O-].O.O.O.O.O

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Related Categories

Application


  • An Analytical Method for Determination of Total Iron in Pharmaceutical-grade Intravenous Iron Colloidal Complexes by Redox-Potentiometry.: This study details a method for determining total iron content in pharmaceutical-grade intravenous iron colloidal complexes using redox-potentiometry. Sodium thiosulfate pentahydrate is utilized in the redox reaction to accurately quantify iron levels. (Kamisetty et al., 2023).

  • New Fluorescent Nanosensor for Determination of Diazepam Using Molecularly Imprinted Mn-doped ZnS Quantum Dots.: This research introduces a new fluorescent nanosensor for detecting diazepam, employing molecularly imprinted Mn-doped ZnS quantum dots. Sodium thiosulfate pentahydrate is used in the synthesis of these quantum dots. (Samadi-Maybodi et al., 2022).

  • Debugging periodate oxidation of cellulose: Why following the common protocol of quenching excess periodate with glycol is a bad idea.: This paper addresses the issues in periodate oxidation of cellulose and suggests alternatives to glycol for quenching excess periodate, including the use of sodium thiosulfate pentahydrate as an efficient quenching agent. (Simon et al., 2023).

  • The effect of sodium thiosulfate on immune cell metabolism during porcine hemorrhage and resuscitation.: Investigates the impact of sodium thiosulfate on immune cell metabolism in a porcine model of hemorrhage and resuscitation, highlighting its potential therapeutic benefits. (Wolfschmitt et al., 2023).

  • Green sulfidated iron oxide nanocomposites for efficient removal of Malachite Green and Rhodamine B from aqueous solution.: This study presents the synthesis of green sulfidated iron oxide nanocomposites using sodium thiosulfate pentahydrate, demonstrating their efficacy in removing dyes like Malachite Green and Rhodamine B from aqueous solutions. (Guo et al., 2022).

Analysis Note

Purity (metallic): ≥ 99.999 %
Assay (iodometric, calculated as pentahydrate): ≥ 99.0 %
Phosphate (PO₄): ≤ 5 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 2.0 ppm
Bi (Bismuth): ≤ 0.01 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 5.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 1.0 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.050 ppm
Ni (Nickel): ≤ 0.05 ppm
Pb (Lead): ≤ 0.05 ppm
Rb (Rubidium): ≤ 1.0 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service