Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06507

Supelco

Sodium dithionite

for analysis EMSURE®

Synonym(s):

Sodium dithionite, Sodium hydrosulfite, Sodium hypodisulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O4S2
CAS Number:
Molecular Weight:
174.11
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-890-0
Assay:
≥85.0% (iodometric)
form:
powder
solubility:
250 g/L (slow decomposition)

Quality Level

product line

EMSURE®

Assay

≥85.0% (iodometric)

form

powder

autoignition temp.

>200 °C

potency

2500 mg/kg LD50, oral (Rat)

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

mp

100 °C (decomposition)

transition temp

flash point >100 °C

solubility

250 g/L (slow decomposition)

density

2.5 g/cm3 at 20 °C

bulk density

1250 kg/m3

anion traces

chloride (Cl-): ≤0.01%

cation traces

Fe: ≤0.002%

storage temp.

2-30°C

InChI

1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2

InChI key

JVBXVOWTABLYPX-UHFFFAOYSA-L

Linkage

Replaces: SX0530; SX0530-1

Analysis Note

Assay (iodometric): ≥ 85 %
Identity: passes test
Chloride (Cl): ≤ 0.01 %
Fe (Iron): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Self-heat. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service