Skip to Content
MilliporeSigma

1.06301

SAFC

CellPrime® rTrypsin

recombinant Trypsin (powder)

Pharma Manufacturing

Synonym(s):

CellPrime® rTrypsin, Parenzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352207
EC Index Number:
232-650-8

Quality Level

form

powder

pH

5 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

200 kg/m3

storage temp.

2-8°C

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Tailored viral vaccine manufacturing addresses virus-specific challenges through collaboration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service