Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06254

Millipore

Neomycin sulfate

for biochemistry

Synonym(s):

Neomycin sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4 · xH2O
CAS Number:
Molecular Weight:
908.88 (anhydrous basis)
UNSPSC Code:
12352201
EC Index Number:
215-773-1

Quality Level

form

solid

pH

5.0-7.5 (20 °C, 50 g/L in H2O)

solubility

500 g/L

bulk density

640 kg/m3

storage temp.

2-8°C

InChI

1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22?,23+;/m1./s1

InChI key

OIXVKQDWLFHVGR-GQTDVWSESA-N

Analysis Note

Identity (IR-spectrum): passes test
pH (50 g/l CO₂-free water): 5.0 - 7.5
Spec. rotation (α 20/D; 100 g/l; water; calc. on dried substance): +53 - +59 °
TLC-Test: passes test
Loss on drying (60 °C; 3 h; 6.7 hPa): ≤ 8 %

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service