Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06161

Supelco

Murexide

metal indicator, ACS

Synonym(s):

Murexide, Ammonium purpurate, acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N6O6
CAS Number:
Molecular Weight:
284.19
MDL number:
UNSPSC Code:
12352107
EC Index Number:
221-266-6
NACRES:
NB.21

product name

Murexide, (ammonium purpurate) metal indicator ACS

grade

ACS reagent

Quality Level

form

solid

loss

≤2.0% loss on drying, 105°C

pH

5 (20 °C, 1 g/L in H2O)

solubility

1 g/L

bulk density

330 kg/m3

Amax

520-524 nm in water

storage temp.

2-30°C

InChI

1S/C8H4N5O6.H3N/c14-3-1(4(15)11-7(18)10-3)9-2-5(16)12-8(19)13-6(2)17;/h(H2,10,11,14,15,18)(H2,12,13,16,17,19);1H3/q-1;/p+1

InChI key

PCCZWUCAYILEMX-UHFFFAOYSA-O

Related Categories

Linkage

Replaces: MX1665-3; MX1665

Analysis Note

Identity (IR-spectrum): passes test
Identity (UV/VIS-Spectrum): passes test
Absorption maximum (water): 520 - 524 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): ≥ 300
Sulfated ash (800 °C): ≤ 1.0 %
Loss on Drying (105°C): ≤ 2.0 %
Suitability as indicator (for metal titration): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service