Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05651

Supelco

ProteoChrom HPTLC plates, cellulose plate

plate L × W 10 cm × 10 cm, suitable for HPTLC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

aluminum support
cellulose matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 25 sheets

technique(s)

HPTLC: suitable

layer thickness

100 μm

plate L × W

10 cm × 10 cm

storage temp.

2-30°C

General description

We’ve enhanced our ProteoChrom plates to make separation highly efficient – especially when peptides and protein digests need to be analyzed. ProteoChrom plates take performance to new levels. Applications are highly reproducible thanks to optimized separation and staining procedures. With extra-thin 100µm layers, the plates are incredibly sensitive. They’re also perfect for use with aqueous solvent systems since they prove highly stable in water. For added convenience, our ProteoChrom plates include easy-to-read and detailed protocols.
for peptide analysis 10 x 10 cm

Analysis Note

Layer thickness: 90 - 120 µm
Deviation of layer thickness per plate: ≤ 20 µm
Aminoacid test
hRf-values
- L-Lysine:
22 - 33
- L-threonine, amino acid test: 38 - 48
- L- Valine: 54 - 66
separation number: ≥ 11
Typical value determined on a sheet
Eluent: 2-Butanol/pyridine/ acetic acid/ water (30/20/6/24 v/v/v/v)

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service