Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.05410

Millipore

Reinforced clostridial agar (RCA)

for microbiology

Synonym(s):

RCM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Quality Level

form

granular

manufacturer/tradename

GranuCult® plus

technique(s)

microbiological culture: suitable

pH

6.7-6.9 (37 °C, 50 g/L in H2O, after autoclaving)

solubility

50 g/L

bulk density

670 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Clostridium spp.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service