Skip to Content
MilliporeSigma
All Photos(1)

Documents

104550

Sigma-Aldrich

ACA

A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets.

Synonym(s):

ACA, N-( p-Amylcinnamoyl)anthranilic Acid, N-(p-Amylcinnamoyl)anthranilic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO3
CAS Number:
Molecular Weight:
337.41
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 10 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C21H23NO3/c1-2-3-4-7-16-10-12-17(13-11-16)14-15-20(23)22-19-9-6-5-8-18(19)21(24)25/h5-6,8-15H,2-4,7H2,1H3,(H,22,23)(H,24,25)/b15-14+

InChI key

GAMRBCZMOOMBSQ-CCEZHUSRSA-N

General description

A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets. Inhibits ATP-stimulated phosphatidylcholine (PC) secretion in alveolar type II cells. Also blocks phorbol ester and A23187-induced PC secretion in type II cells. Reported to block glucose-induced insulin secretion by pancreatic β-cells. Exhibits moderate leukotriene antagonist activity.
Inhibits epinephrine-stimulated thromboxane production (86% at 3.5 μM) via inhibition of phospholipase A2 (PLA2) in human platelets. Also blocks glucose-induced insulin secretion in pancreatic islets. Possesses moderate leukotriene antagonist activity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
A2PLA2
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Liu, L., et al. 1999. J. Cell. Biochem.72, 103.
Simonsson, E., et al. 1998. Diabetes47, 1436.
Konrad, R.J., et al. 1992. Biochim. Biophys. Acta1135, 215.
Banga, H.S., et al. 1986. Proc. Natl. Acad. Sci. USA83, 9197.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service