Recommended Products
form
solid
Quality Level
potency
>5000 mg/kg LD50, oral (Rat)
composition
Dye, ≥60% spectrophotometric assay
pH
5.0 (20 °C, 10 g/L in H2O)
solubility
70 g/L
bulk density
670 kg/m3
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/C37H29N3O9S3.2Na/c41-50(42,43)34-10-4-1-7-31(34)38-28-19-13-25(14-20-28)37(26-15-21-29(22-16-26)39-32-8-2-5-11-35(32)51(44,45)46)27-17-23-30(24-18-27)40-33-9-3-6-12-36(33)52(47,48)49;;/h1-24,38-39H,(H,41,42,43)(H,44,45,46)(H,47,48,49);;/q;2*+1/p-3
InChI key
TUHAIJABPUJAEY-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (Hydrochloric acid 0.005 mol/l): 595 - 605 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; hydrochloric acid 0.005 mol/l): 675 - 1125
TLC-Test: passes test
Loss on drying (110 °C): ≤ 7 %
Suitability for microscopy: passes test
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
572.0 °F - Pensky-Martens closed cup
Flash Point(C)
> 300 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service