Skip to Content
MilliporeSigma

1.04078

SAFC

Gelatin

powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, NF

Pharma Manufacturing

Synonym(s):

Gelatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
EC Index Number:
232-554-6

Agency

BP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

coarse powder
powder

potency

>5000 mg/kg LD50, oral (Rat)

pH

3.8-7.6 (25 °C, 10 g/L in H2O)

application(s)

pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Gelatin is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical application, gelatin has been used primarily for solid oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Application

Product No.
Description
Pricing

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service