Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04013

Supelco

Furfural

GR for analysis ACS

Synonym(s):

Furfural, 2-Furancarbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
MDL number:
UNSPSC Code:
12352114
EC Index Number:
202-627-7

grade

ACS reagent

vapor pressure

1 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

315 °C

potency

65 mg/kg LD50, oral (Rat)

expl. lim.

2.1-19.3 % (v/v)

bp

162 °C/1013 hPa

mp

-37 °C

transition temp

flash point 60 °C

solubility

83 g/L

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H

InChI key

HYBBIBNJHNGZAN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Density (d 20 °C/20 °C): 1.155 - 1.163
Titrable free acid: ≤ 0.02 meq/g
Evaporation residue: ≤ 0.5 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service