Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03956

Supelco

Iron(II) sulfide

sticks Ø ~ 1 cm

Synonym(s):

Iron(II) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeS
CAS Number:
Molecular Weight:
87.91
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-268-6
NACRES:
NB.51

Quality Level

form

solid

concentration

~25% (sulfide)

mp

1195 °C

density

4.84 g/cm3 at 20 °C

bulk density

1500‑2000 kg/m3

storage temp.

no temp limit

InChI

1S/Fe.S

InChI key

MBMLMWLHJBBADN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Role of iron(II) sulfide in autotrophic denitrification under tetracycline stress: Substrate and detoxification effect.: This study investigates the role of Iron(II) sulfide in enhancing autotrophic denitrification processes under the stress of tetracycline, revealing its potential for wastewater treatment applications. It highlights how FeS can act as both a substrate and a detoxification agent, thus improving the efficiency of bioremediation efforts (Bai et al., 2022).

  • Speciation of iron sulfide compounds by means of X-ray Emission Spectroscopy using a compact full-cylinder von Hamos spectrometer.: This paper details a novel application of X-ray Emission Spectroscopy (XES) to study iron sulfide compounds. The study utilizes a compact full-cylinder von Hamos spectrometer, providing insights into the speciation of iron sulfides, which is critical for understanding their chemical behavior and potential industrial applications (Wansleben et al., 2020).

  • A novel electrochemical sensor based on FeS anchored reduced graphene oxide nanosheets for simultaneous determination of dopamine and acetaminophen.: This research introduces an innovative electrochemical sensor using Iron(II) sulfide anchored on reduced graphene oxide nanosheets. The sensor is designed for the simultaneous detection of dopamine and acetaminophen, showcasing the potential of FeS in the development of sensitive and selective biosensors for medical diagnostics (Liu et al., 2017).

Analysis Note

Assay (sulphide): about 25 %
Identity: passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service