Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.03424

Sigma-Aldrich

Dimethyl sulfoxide-d6

deuteration degree min. 99.8% for NMR spectroscopy MagniSolv

Synonym(s):

DMSO deuterated, (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 0.5 ML
$65.30
10 X 0.75 ML
$85.20
10 ML
$98.70
10 ML
$103.00
25 ML
$196.00
50 ML
$328.00
100 ML
$582.00

$65.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 X 0.5 ML
$65.30
10 X 0.75 ML
$85.20
10 ML
$98.70
10 ML
$103.00
25 ML
$196.00
50 ML
$328.00
100 ML
$582.00

About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
MDL number:
UNSPSC Code:
12142201
EC Index Number:
218-617-0
NACRES:
NA.22

$65.30


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

form

liquid

autoignition temp.

573 °F

potency

14500 mg/kg LD50, oral (Rat)
40000 mg/kg LD50, skin (Rat)

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.020% Water (H<SUB>2</SUB>O) (NMR)
≤0.030% Water (H<SUB>2</SUB>O + D<SUB>2</SUB>O) (Karl Fischer)

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C

transition temp

flash point 88 °C

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

storage temp.

no temp limit

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Computer modeling of adsorption on an activated charcoal surface.: This study uses computational modeling to simulate adsorption processes, where Dimethyl sulfoxide-d₆ is likely used as a model solvent to analyze interactions at the molecular level (Wurster et al., 1994).

  • Solvent effects on the conformation of cyclo(-D-Trp-D-Asp-Pro-D-Val-Leu-). An NMR spectroscopy and molecular modeling study.: Investigates how solvents like Dimethyl sulfoxide-d₆ influence peptide conformations, enhancing our understanding of peptide behavior in different chemical environments (Gonnella et al., 1994).

  • Role of the NH2-terminal domain of angiotensin II (ANG II) and [Sar1]angiotensin II on conformation and activity. NMR evidence for aromatic ring clustering and peptide backbone folding compared with [des-1,2,3]angiotensin II.: Discusses the structural and functional aspects of angiotensin II with Dimethyl sulfoxide-d₆ as a solvent to facilitate NMR analysis, providing insights into peptide structure-activity relationships (Matsoukas et al., 1994).

  • Cyclic lactam analogues of Ac-[Nle4]alpha-MSH4-11-NH2.: Focuses on cyclic lactam analogues using Dimethyl sulfoxide-d₆ in NMR studies to investigate molecular conformations, contributing to peptide chemistry research (Sugg et al., 1988).

  • NMR study of a lymphocyte differentiating thymic factor. An investigation of the Zn(II)-nonapeptide complexes (thymulin).: Utilizes Dimethyl sulfoxide-d₆ in detailed NMR studies to explore the interactions of metal-peptide complexes, which are crucial for understanding the bioactive conformation and function of peptides in analytical biochemistry (Cung et al., 1988).

Analysis Note

Deuteration degree: ≥ 99.80 %
Water (K. Fischer, H₂O + D₂O): ≤ 0.030 %
Performance test (NMR-spectrum): passes test
Water (NMR, H₂O): ≤ 0.020 %

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service