Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03121

Supelco

N,N-Diethyl-1,4-phenylenediammonium sulfate

GR for analysis

Synonym(s):

N,N-Diethyl-1,4-phenylenediammonium sulfate, 4-Amino-N,N-diethylaniline sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$194.00
25 KG
$42,900.00

$194.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$194.00
25 KG
$42,900.00

About This Item

Empirical Formula (Hill Notation):
C10H18N2O4S
CAS Number:
Molecular Weight:
262.33
MDL number:
UNSPSC Code:
12352107
EC Index Number:
228-500-6
NACRES:
NA.05

$194.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

autoignition temp.

540 °C

potency

497 mg/kg LD50, oral (Rat)

pH

2.0-2.2 (20 °C, 50 g/L in H2O)

bp

274-275 °C/1013 hPa

mp

184-186 °C

solubility

500 g/L

bulk density

580 kg/m3

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+H](CC)(CC)c1ccc(cc1)[N+H3]

InChI

1S/C10H16N2.H2O4S/c1-3-12(4-2)10-7-5-9(11)6-8-10;1-5(2,3)4/h5-8H,3-4,11H2,1-2H3;(H2,1,2,3,4)

InChI key

AYLDJQABCMPYEN-UHFFFAOYSA-N

Analysis Note

Assay: ≥ 99.5 %
Identity (IR-spectrum): passes test
pH-value (5 %; water): 2.0 - 2.2
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Sulfated ash (600 °C): ≤ 0.05 %
Water (according to Karl Fischer): ≤ 0.5 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service