Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03110

Supelco

1.4-Dioxane

dried SeccoSolv

Synonym(s):

1.4-Dioxane, Glycolethylether, 1,4-Diethylene dioxide, 1,4-Dioxacyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8

vapor pressure

41 hPa ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

300 °C

potency

5200 mg/kg LD50, oral (Rat)
7600 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-25.2 % (v/v)

technique(s)

DNA amplification: suitable
titration: suitable

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.005% Water
≤0.0050% Peroxide (as H2O2)

evapn. residue

≤10 mg/L

color

APHA: ≤10

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Peroxide (as H₂O₂): ≤ 0.0050 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.005 %
The product is stabilised with about 2,5ppm 2,6-Di-tert-butyl-4-methylphenol (BHT).

Legal Information

SECCOSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service