Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03037

Supelco

2,6-Dichloroquinone-4-chloroimide

GR for analysis reagent for Vitamin B₆ Reag. Ph Eur

Synonym(s):

2,6-Dichloroquinone-4-chloroimide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$751.00

$751.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$751.00

About This Item

Empirical Formula (Hill Notation):
C6H2Cl3NO
CAS Number:
Molecular Weight:
210.45
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-937-2
NACRES:
NA.21

$751.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

solid

mp

65-67 °C

bulk density

800 kg/m3

storage temp.

2-8°C

SMILES string

ClN=C1C=C(C(=O)C(=C1)Cl)Cl

InChI

1S/C6H2Cl3NO/c7-4-1-3(10-9)2-5(8)6(4)11/h1-2H

InChI key

YHUMTHWQGWPJOQ-UHFFFAOYSA-N

Application


  • Utilizing experimental design and desirability function in optimizing RP-HPLC method: Discusses the optimization of analytical methods using 30% acetic acid, improving the detection and analysis of pharmaceutical compounds (Mozayad et al., 2024).

  • Combined application of bacteriophages with a competitive exclusion culture and carvacrol with organic acids can reduce Campylobacter in primary broiler production: Illustrates the role of 30% acetic acid in agricultural biocontrol applications, reducing pathogen load in poultry (Peh et al., 2024).


Analysis Note

Assay (iodometric): ≥ 98
Identity (IR-spectrum): passes test
Spec. Absorptivity A 1%/1cm (312 nm; 0,001 %; 1 cm; ethanol): ≥ 800
Melting point: 65 - 68

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service