Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02836

SAFC

L-Cystine

suitable for biopharmaceutical production FCC

Synonym(s):

L-Cystine, Cys

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-296-3

reg. compliance

FCC

Quality Level

form

solid

mp

261-262 °C (decomposition)

solubility

insoluble 0.19 g/L

density

1.677 g/cm3 at 20 °C

bulk density

300 kg/m3

application(s)

pharma/biopharma processes

storage temp.

2-30°C

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Analysis Note

Assay (ex N, calc. on dried substance): 98.5 - 101.5 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine-crystalline powder
Appearance of solution (100 g/l, hydrochloric acid  2 mol/l): clear and colorless
Spec. rotation (α 20/D, 20 g/l, hydrochloric acid 1 mol/l, previously dried substance): -215 to -225
Heavy metals (as Pb): ≤ 10 ppm
Pb (Lead): ≤ 5 ppm
Ninhydrin - positive substances (LC) (largest single impurity): ≤ 0.2 %
Ninhydrin - positive substances (LC) (Sum of all impurities): ≤ 0.5 %
Sulfated ash (800 °C): ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.2 %
Corresponds to FCC V

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service