Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02546

SAFC

Cobalt(II) sulfate heptahydrate

extra pure

Synonym(s):

Cobalt(II) sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
281.10
UNSPSC Code:
12352107
EC Index Number:
233-334-2

Quality Level

form

solid

potency

582 mg/kg LD50, oral (Rat)

pH

4 (20 °C, 100 g/L in H2O)

mp

98 °C

solubility

362 g/L

density

1.95 g/cm3 at 20 °C

bulk density

900 kg/m3

application(s)

pharma/biopharma processes

storage temp.

15-25°C

InChI

1S/Co.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

MEYVLGVRTYSQHI-UHFFFAOYSA-L

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Cobalt (II) sulfate heptahydrate is used as reagent, as source of cobalt.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service