Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02414

Sigma-Aldrich

Warthin-Starry silver plating kit, modified

for bacteria: Campylobacter pylori

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124

IVD

for in vitro diagnostic use

Quality Level

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

suitability

bacteria: Campylobacter pylori (Helicobacter pylori)

General description

Warthin-Starry silver plating kit modified - for the detection of Helicobacter pylori and Spirochetes in paraffin sections, is used to specifically detect the bacterium Helicobacter pylori in histological specimens of human origin. The bacteria are found in the mucus of the surface epithelium, in the apical gastric glands and in the veolae of the gastric mucosa. They appear dark brown to black in the microscopic image, whereas surrounding cells are stained yellow and the nuclei brown. The kit contains four reagents: 6% Silver nitrate solution, Hydroquinone mixture, Gelatine powder and 1.2% Acetic acid solution, which are ready-to-use. To prevent fading, the staining preparations need to be cover-slipped with DPX new (Product number 1.00579) or Neo-Mount(Product number 1.09016); an alternative xylene-containing covering agent (Xylene (isometric, Product number 1.08298) can be used if an additional silver-fixing step with sodium thiosulfate is included previous covering.
The kit is sufficient for up to 500 preparations. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

For the detection of Helicobacter pylori and Spirochetes in paraffin sections

Legal Information

NEO-MOUNT is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service