Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02284

Supelco

Quinhydrone

GR for analysis

Synonym(s):

Quinhydrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O4
CAS Number:
Molecular Weight:
218.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-387-6

Quality Level

form

solid

potency

225 mg/kg LD50, oral (Rat)

solubility

4 g/L

density

1.40 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-30°C

InChI

1S/C6H6O2.C6H4O2/c2*7-5-1-2-6(8)4-3-5/h1-4,7-8H;1-4H

InChI key

BDJXVNRFAQSMAA-UHFFFAOYSA-N

Analysis Note

Assay p-Benzoquinone: 48 - 52
assay hydroquinone: 48 - 52
Identity (IR-spectrum): passes test
Matter insoluble in ethanol: ≤ 0.02 %
Sulphate (SO₄): ≤ 0.01 %
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service