Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02282

Supelco

Quinaldine red

pH indicator, Reag. Ph Eur

Synonym(s):

Quinaldine red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23IN2
CAS Number:
Molecular Weight:
430.33
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-221-5
NACRES:
NA.21

product name

Quinaldine red, indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

impurities

≤5% Water (Karl Fischer)

visual transition interval

1.4-3.2, colorless to red

mp

240 °C (decomposition)

bulk density

290 kg/m3

λmax

528-533 nm in acetic acid

storage temp.

2-30°C

InChI

1S/C21H23N2.HI/c1-4-23-20(16-12-18-7-5-6-8-21(18)23)15-11-17-9-13-19(14-10-17)22(2)3;/h5-16H,4H2,1-3H3;1H/q+1;/p-1

InChI key

JOLANDVPGMEGLK-UHFFFAOYSA-M

Related Categories

Application


  • Quinaldine Red as a fluorescent probe for determining the melting temperature (T(m)) of proteins: a simple, rapid and high-throughput assay.: This study presents Quinaldine Red as an effective fluorescent probe for determining the melting temperatures of proteins, offering a rapid and high-throughput method suitable for broad applications in biochemical research (Das et al., 2024).

  • Red emitting fluorogenic dye as an efficient turn-on probe for milk allergen.: Explores the use of a red-emitting fluorogenic dye, likely involving Quinaldine Red, as a turn-on probe for detecting milk allergens, highlighting its potential in food safety and allergen detection (Chakraborty, 2022).

  • A rapid, naked-eye detection of hypochlorite and bisulfite using a robust and highly-photostable indicator dye Quinaldine Red in aqueous medium.: Describes the application of Quinaldine Red for the visual detection of chemicals like hypochlorite and bisulfite, underlining its utility in environmental monitoring and chemical safety (Dutta et al., 2018).

  • Self-Assembly and Formation of Chromonic Liquid Crystals from the Dyes Quinaldine Red Acetate and Pyronin Y.: Investigates the self-assembly properties of Quinaldine Red and its application in forming chromonic liquid crystals, demonstrating its potential in material science and nanotechnology (Magana et al., 2016).

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Transition range: pH 1.4 - pH 3.2 colourless - red
Absorption maximum λmax. (Acetic acid): 528 - 533 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; acetic acid glacial; calculated on anhydrous substance): 1150 - 1480
Water (according to Karl Fischer): ≤ 5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service