Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02103

Supelco

Calcium lactate

for soil tests

Synonym(s):

Calcium lactate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
308.29
MDL number:
UNSPSC Code:
12352107
EC Index Number:
248-953-3

Quality Level

autoignition temp.

610 °C

potency

3730 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

7 (20 °C, 50 g/L in H2O)

mp

240 °C

solubility

50 g/L

bulk density

300‑500 kg/m3

storage temp.

15-25°C

InChI

1S/2C3H6O3.Ca.5H2O/c2*1-2(4)3(5)6;;;;;;/h2*2,4H,1H3,(H,5,6);;5*1H2/q;;+2;;;;;/p-2

InChI key

HPVJXNNKHRNBOY-UHFFFAOYSA-L

General description

for soil tests

Analysis Note

Assay (complexometric, calc. on anhydrous substance)≥ 98 %Identity (IR-spectrum)passes testpH-value (5 %; water)6 - 7K (Potassium)≤ 0.01 %Phosphate (PO₄)≤ 0.002 %Water (according to Karl Fischer)22 - 30 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service