Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.02067

Supelco

Calcium carbonate

precipitated for analysis of silicates EMSURE®

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$1,530.00

$1,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$1,530.00

About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-439-9
NACRES:
NA.21
form:
solid
solubility:
0.017 g/L

$1,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

6450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

impurities

≤0.005% Substances insoluble in hydrochloric acid

pH

9.5-10.5 (20 °C, 100 g/L in H2O, slurry)

mp

825 °C (decomposition)

solubility

0.017 g/L

density

2.93 g/mL at 25 °C (lit.)

bulk density

300‑1400 kg/m3

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.02%
Fe: ≤0.001%
K: ≤0.002%
Mg: ≤0.02%
N: ≤0.01%
Na: ≤0.005%
Sr: ≤0.02%
heavy metals (as Pb): ≤0.001%

storage temp.

10-30°C

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization of an advanced bone graft material with a nanocrystalline hydroxycarbanoapatite surface and dual phase composition.: The study characterizes an advanced bone graft material, emphasizing the role of calcium carbonate in enhancing the material′s structural and functional properties for biomedical applications (Borden et al., 2024).

  • Discovery of a hyperalkaline liquid condensed phase: significance toward applications in carbon dioxide sequestration.: This study explores a hyperalkaline liquid phase rich in calcium carbonate, discussing its potential applications in carbon dioxide sequestration and environmental remediation (Bewernitz et al., 2024).


Analysis Note

Assay (complexometric): ≥ 99.0 %
Substances insoluble in hydrochloric acid: ≤ 0.005 %
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Total nitrogen (N): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
Ba (Barium): ≤ 0.02 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.02 %
Na (Sodium): ≤ 0.005 %
Sr (Strontium): ≤ 0.02 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service