Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02037

Supelco

Cesium chloride-aluminium nitrate buffer solution

acc. to Schuhknecht and Schinkel for the determination of sodium, potassium and lithium by flame spectrometry (enth./cont. 50 g/l CsCl + 250 g/l Al(NO₃)₃ * 9 H₂O)

Synonym(s):

CsCl-Al(NO3)3 Mixed Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

form

liquid

storage condition

(Tightly closed. Keep locked up or in an area accessible only to qualified or authorized persons.)

concentration

( Aluminium nitrate: >= 10 - < 20% Cesium chloride : >= 1 - < 5%)

technique(s)

AAS: suitable

color

colorless

pH

2.5 (20 °C in H2O)

solubility

soluble
water: soluble

density

1.14 g/cm3 at 20 °C

cation traces

K: ≤1 mg/L
Li: ≤1 mg/L
Na: ≤5 mg/L

storage temp.

15-25°C

Analysis Note

Content of Aluminiumnitrat-nonahydrat (potentiometric): 247.5 - 252.5 g/l
Content of Cesiumchlorid (potentiometric): 49.5 - 50.5 g/l
Li (Lithium): <1 mg/l
K (Potassium): <1 mg/l
Na (Sodium): <5 mg/l

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service