Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01789

Sigma-Aldrich

Benzene-D6

deuteration degree min. 99.5% for NMR spectroscopy MagniSolv

Synonym(s):

Benzene-D6, Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
MDL number:
UNSPSC Code:
12142201
EC Index Number:
214-061-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

101 hPa ( 20 °C)

Quality Level

isotopic purity

≥99.5 atom % D

form

liquid

autoignition temp.

555 °C

potency

930 mg/kg LD50, oral (Rat)
>8260 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-8.0 % (v/v)

technique(s)

NMR: suitable

impurities

≤0.02% Water (HO) (NMR)

kinematic viscosity

0.78 cSt(20 °C)

bp

79 °C/1013 hPa

mp

6.7 °C

transition temp

flash point -11 °C

density

0.95 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

c1(c(c(c(c(c1[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Application

Benzene-D6 deuteration degree min. 99.5% for NMR spectroscopy MagniSolv. CAS 1076-43-3, molar mass 84.15 g/mol.

Analysis Note

Deuteration degree: ≥ 99.5 %
Water (NMR, HO): ≤ 0.02 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service