Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01771

Supelco

FAM Benzine

DIN 51635

Synonym(s):

Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352401
bp:
65 °C/1013 hPa
vapor pressure:
10 hPa ( 20 °C)

vapor pressure

10 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

245 °C

potency

>5840 mg/kg LD50, oral (Rat)

expl. lim.

0.7-8.3 % (v/v)

impurities

≤0.002% Non volatile matter
≤0.1% Aromatics (as benzene)
≤5% n-Hexane (GC)

non-volatile residue (NVR)

≤0.002%

color

APHA: ≤10

bp

65 °C/1013 hPa

mp

<-30 °C

transition temp

flash point -26 °C

solubility

slightly soluble

density

0.76 g/cm3 at 15 °C

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Colour: ≤ 10 Hazen
Density (15°C/DIN 51757): 0.690 - 0.710
Boiling range (DIN 51751 (60-90°C)): 85 - 95 % (v/v)
Aniline point (DIN ISO 2977): 59.0 - 61.0 °C
Aromatics (as benzene): ≤ 0.1 %
n-Hexane (GC): ≤ 5.0 %
Non volatile matter: ≤ 0.002 %

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service