Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01749

Supelco

Volatile organic Acids Test, photometric

photometric, 50-3000 mg/L (acetic acid), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
$170.00

$170.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
$170.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$170.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Volatile organic Acids Cell Test, photometric, 50-3000 mg/L (acetic acid), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

analyte functional class(es)

volatiles 

analyte chemical class(es)

organic acids

measuring range

50-3000 mg/L (acetic acid)
50-3000 mg/L (butyric acid)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Hydroxamic acids / iron(III) salt)

storage temp.

15-25°C

General description

Method: photometric 50 - 3000 mg/l Spectroquant®

This Spectroquant® Volatile Organic Acids Cell Test allows the accurate determination of the concentration of lower fatty acids (“volatile organic acids”) in aqueous samples.

Method applied: In an acidic medium, lower fatty acids react with a diole to form fatty acid esters, which are subsequently converted into hydroxamic acids with hydroxylamine. These in turn react with iron(III) ions to form red complexes that are determined photometrically.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service